Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010722.5 | NP_034852.2 | 5 | 766 | |
GenBank mRNA | AK132671.1 | - | 2 | 474 | |
BC042430.1 | - | 5 | 771 | 3674002 | |
BC051985.1 | - | 5 | 742 | 5695459 | |
D13455.1 | - | 2 | 453 | ||
X54098.1 | - | 5 | 725 |