Gene Transcripts

Gene Symbol
Sln
Entrez Gene ID
367086
Gene Name
sarcolipin
Gene Aliases
Chromosome Location
Chr. 8: 58431407 - 58435654 on Build Rnor_6.0
UniGene ID
Rn.16457
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013247.1 NP_001013265.1 2 178
GenBank mRNA AY456000.1 - 2 178

View Full Product Details