Gene Transcripts
- Gene Symbol
- CAMK2B
- Entrez Gene ID
- 816
- Gene Name
- calcium/calmodulin dependent protein kinase II beta
- Gene Aliases
- CAM2, CAMK2, CAMKB
- Chromosome Location
- Chr. 7: 44217150 - 44325994 on Build GRCh38
- UniGene ID
- Hs.351887
- Species
- Homo sapiens
View Full Product Details