Gene Transcripts

Gene Symbol
FRRS1L
Entrez Gene ID
23732
Gene Name
ferric chelate reductase 1 like
Gene Aliases
C9orf4, CG-6, CG6, EIEE37
Chromosome Location
Chr. 9: 109136188 - 109167291 on Build GRCh38
UniGene ID
Hs.347537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014334.2 NP_055149.2 3 507
XM_011518453.2 XP_011516755.1 3 320
XM_011518454.2 XP_011516756.1 4 891
GenBank mRNA AF155065.1 - 3 507
BF514667.1 - 3 348

View Full Product Details