Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199579.1 | NP_001186508.1 | 16 | 2270 | |
NM_001199580.1 | NP_001186509.1 | 17 | 2206 | ||
NM_015374.2 | NP_056189.1 | 16 | 2207 | ||
XM_011530104.2 | XP_011528406.1 | 15 | 2064 | ||
XM_011530105.2 | XP_011528407.1 | 16 | 1931 | ||
XM_017028748.1 | XP_016884237.1 | 16 | 2202 | ||
GenBank mRNA | AB014568.1 | - | 16 | 1989 | |
AF202723.1 | - | 8 | 1082 | ||
AK295787.1 | - | 16 | 1931 | ||
AK304188.1 | - | 17 | 2066 | ||
AY682988.1 | - | 16 | 1988 | ||
BC030684.2 | - | 4 | 412 | ||
BC069253.1 | - | 8 | 1088 | ||
BC094797.1 | - | 16 | 2227 | 4799090 | |
BC111549.1 | - | 15 | 1936 | 40008751 | |
BC111717.1 | - | 15 | 1936 | 40008756 | |
BX537962.1 | - | 17 | 2198 | ||
CR456474.1 | - | 15 | 1934 |