Gene Transcripts
- Gene Symbol
- ZZZ3
- Entrez Gene ID
- 26009
- Gene Name
- zinc finger ZZ-type containing 3
- Gene Aliases
- ATAC1
- Chromosome Location
- Chr. 1: 77561526 - 77683440 on Build GRCh38
- UniGene ID
- Hs.480506
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308237.1 |
NP_001295166.1
|
7, 8 |
916 |
|
NM_015534.5 |
NP_056349.1
|
8, 9 |
2380 |
|
XM_005270725.3 |
XP_005270782.1
|
8, 9 |
2455 |
|
XM_005270726.3 |
XP_005270783.1
|
7, 8 |
2356 |
|
XM_005270727.3 |
XP_005270784.1
|
8, 9 |
2452 |
|
XM_005270729.4 |
XP_005270786.1
|
6, 7 |
866 |
|
XM_017000970.1 |
XP_016856459.1
|
7, 8 |
2353 |
|
XM_017000972.1 |
XP_016856461.1
|
7, 8 |
884 |
|
GenBank mRNA |
AK074119.1 |
- |
4, 5 |
1426 |
|
BC035079.2 |
- |
7, 8 |
908 |
|
BC035397.1 |
- |
4, 5 |
1048 |
|
BC035818.2 |
- |
4, 5 |
1955 |
5554765 |
BX640658.1 |
- |
4, 5 |
468 |
|
BX640766.1 |
- |
4, 5 |
1654 |
|
BX641001.1 |
- |
8, 9 |
2381 |
|
BX641096.1 |
- |
8, 9 |
2366 |
|
View Full Product Details