Gene Transcripts

Gene Symbol
PTPN18
Entrez Gene ID
26469
Gene Name
protein tyrosine phosphatase, non-receptor type 18
Gene Aliases
BDP1, PTP-HSCF
Chromosome Location
Chr. 2: 130356007 - 130375409 on Build GRCh38
UniGene ID
Hs.516390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014369.3 NP_055184.2 2 282
XM_006712417.1 XP_006712480.1 2 277
XM_017003798.1 XP_016859287.1 2 233
GenBank mRNA DN995571.1 - 2 290
X79568.1 - 2 224

View Full Product Details