Gene Transcripts

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
Chromosome Location
Chr. 10: 96189171 - 96271576 on Build GRCh38
UniGene ID
Hs.665244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114094.1 NP_001107566.1 5 424
NM_001258440.1 NP_001245369.1 5 364
NM_001258441.1 NP_001245370.1 5 364
NM_001258442.1 NP_001245371.1 5 364
NM_013314.3 NP_037446.1 5 424
NR_047680.1 5 364
NR_047681.1 5 369
NR_047682.1 5 364
NR_047683.1 5 364
XM_011539728.2 XP_011538030.1 5 301
XM_017016159.1 XP_016871648.1 5 302
GenBank mRNA AF068180.1 - 5 399
AF068181.1 - 5 399
AK027779.1 - 5 359
AK225546.1 - 5 407
AM180327.1 - 5 246
AM180328.1 - 5 246
AM180329.1 - 5 246
AM180330.1 - 5 246
AM180331.1 - 5 246
AM180332.1 - 5 246
AM180333.1 - 5 246
AM180334.1 - 5 246
AM180336.1 - 5 246
AM180337.1 - 5 246
AM180338.1 - 5 251
AM180339.1 - 5 246
AM180340.1 - 5 246
AM180341.1 - 4 196
AM180342.1 - 4 196
AM180343.1 - 4 196
AM180344.1 - 3 155
AM180345.1 - 4 196
AM180346.1 - 3 194
BC018906.2 - 5 338 4053894
BC110298.1 - 5 356
BM820310.1 - 5 417
BU597768.1 - 5 369
DA045252.1 - 5 424

View Full Product Details