Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
Chromosome Location
Chr. 6: 143059363 - 143343594 on Build GRCh38
UniGene ID
Hs.567501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286587.1 NP_001273516.1 6 577
NM_016108.3 NP_057192.2 5 634
XM_011535881.2 XP_011534183.1 4 500
XM_011535882.2 XP_011534184.1 4 499
XM_011535883.2 XP_011534185.1 5 615
XM_011535884.2 XP_011534186.1 5 617
XM_011535885.2 XP_011534187.1 4 499
XM_017010927.1 XP_016866416.1 5 633
XM_017010928.1 XP_016866417.1 4 499
GenBank mRNA AF153605.1 - 5 577
AJ420480.1 - 3 250
AK001347.1 - 5 575
AK222475.1 - 5 596
BC025278.1 - 5 565 4859216
BX538067.1 - 4 337

View Full Product Details