Gene Transcripts

Gene Symbol
STK26
Entrez Gene ID
51765
Gene Name
serine/threonine protein kinase 26
Gene Aliases
MASK, MST4
Chromosome Location
Chr. X: 132023217 - 132075943 on Build GRCh38
UniGene ID
Hs.444247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042452.1 NP_001035917.1 6 693
NM_001042453.1 NP_001035918.1 7 939
NM_016542.3 NP_057626.2 8 1170
XM_011531349.1 XP_011529651.1 9 1143
XM_011531350.1 XP_011529652.1 8 1216
GenBank mRNA AB040057.2 - 8 1089
AF231012.1 - 8 1058
AF344882.1 - 7 879
AF344883.1 - 6 693
AK000556.1 - 4 502
AK027589.1 - 3 387
AK074837.1 - 3 423
AK075107.1 - 7 851
AK075150.1 - 8 1085
AK303583.1 - 9 1151
AK314356.1 - 8 1086
AK316137.1 - 9 1151
BC098315.1 - 8 1075 40026349
BC099843.1 - 8 1075 40026348
BC103503.1 - 8 1075 40026350
BT020099.1 - 7 869

View Full Product Details