Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018011.3 | NP_060481.3 | 2 | 705 | |
GenBank mRNA | AK001016.1 | - | 2 | 703 | |
AK303575.1 | - | 3 | 478 | ||
AK304751.1 | - | 3 | 535 | ||
AK309941.1 | - | 2 | 324 | ||
BC050434.1 | - | 2 | 652 | 6016552 | |
BC071587.1 | - | 2 | 676 | 5271043 |