Gene Transcripts

Gene Symbol
SMG8
Entrez Gene ID
55181
Gene Name
SMG8, nonsense mediated mRNA decay factor
Gene Aliases
C17orf71
Chromosome Location
Chr. 17: 59210010 - 59215250 on Build GRCh38
UniGene ID
Hs.7296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018149.6 NP_060619.4 1 1268
GenBank mRNA AF349467.1 - 1 1268
AK001449.1 - 1 417
AL834490.1 - 1 1247
BC020957.1 - 1 436 3851898
BC031604.1 - 1 1214

View Full Product Details