Gene Transcripts

Gene Symbol
ANKRD10
Entrez Gene ID
55608
Gene Name
ankyrin repeat domain 10
Gene Aliases
Chromosome Location
Chr. 13: 110878540 - 110915107 on Build GRCh38
UniGene ID
Hs.525163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017664.3 NP_060134.2 5 937
GenBank mRNA AK000100.1 - 5 899
AK026017.1 - 11
AK094222.1 - 2 937
AK095763.1 - 3 464

View Full Product Details