Gene Transcripts

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
Chromosome Location
Chr. 12: 26905171 - 26938326 on Build GRCh38
UniGene ID
Hs.505077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018164.2 NP_060634.2 16 2496
XM_005253422.3 XP_005253479.1 15 2284
XM_011520750.2 XP_011519052.1 16 2047
XM_011520751.2 XP_011519053.1 15 2302
XM_011520752.2 XP_011519054.1 15 2248
XM_011520753.2 XP_011519055.1 10 1219
XM_017019632.1 XP_016875121.1 16 2029
XM_017019633.1 XP_016875122.1 15 2439
XM_017019634.1 XP_016875123.1 15 2421
XM_017019635.1 XP_016875124.1 15 1858
XM_017019636.1 XP_016875125.1 15 2230
XM_017019637.1 XP_016875126.1 14 1659
GenBank mRNA AF274950.1 - 7 908
AK001222.1 - 6 811
AK001492.1 - 16 2125
AK001499.1 - 16 2161
AK225336.1 - 16 2147
AK297948.1 - 14 1836
AL137401.1 - 3 259
BC003081.2 - 16 2185 2989678
BC008368.1 - 16 2143 3355282

View Full Product Details