Gene Transcripts
- Gene Symbol
- C16orf62
- Entrez Gene ID
- 57020
- Gene Name
- chromosome 16 open reading frame 62
- Gene Aliases
- Chromosome Location
- Chr. 16: 19555415 - 19701163 on Build GRCh38
- UniGene ID
- Hs.654964
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300743.1 |
NP_001287672.1
|
4, 5 |
715 |
|
NM_020314.5 |
NP_064710.4
|
4, 5 |
715 |
|
XM_005255435.3 |
XP_005255492.1
|
4, 5 |
667 |
|
XM_006721065.3 |
XP_006721128.1
|
4, 5 |
667 |
|
XM_011545896.2 |
XP_011544198.1
|
3, 4 |
345 |
|
GenBank mRNA |
AK095078.1 |
- |
4, 5 |
412 |
|
AK290286.1 |
- |
4, 5 |
412 |
|
AK294885.1 |
- |
4, 5 |
715 |
|
AL832159.1 |
- |
4, 5 |
411 |
|
AL833428.1 |
- |
4, 5 |
404 |
|
BC014900.2 |
- |
3, 4 |
383 |
3909068 |
BC050464.1 |
- |
4, 5 |
455 |
6061139 |
BC058845.1 |
- |
4, 5 |
414 |
6452778 |
BX537867.1 |
- |
4, 5 |
427 |
|
View Full Product Details