Gene Transcripts

Gene Symbol
RSRP1
Entrez Gene ID
57035
Gene Name
arginine and serine rich protein 1
Gene Aliases
C1orf63, NPD014
Chromosome Location
Chr. 1: 25242237 - 25338254 on Build GRCh38
UniGene ID
Hs.259412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321772.1 NP_001308701.1 4 2026
NM_020317.4 NP_064713.3 4 1022
NR_135143.1 3 2538
NR_135144.1 4 1613
NR_135777.1 3 2513
NR_135778.1 4 1887
NR_135780.1 4 1991
NR_135781.1 5 1658
NR_135782.1 5 1340
NR_135783.1 5 1067
NR_135784.1 3 2538
NR_135785.1 5 1066
NR_135786.1 3 2538
NR_135787.1 5 2662
NR_135788.1 3 2604
NR_135789.1 3 3542
XM_011541797.1 XP_011540099.1 4 987
GenBank mRNA AF267856.1 - 3 2466
AK226012.1 - 4 1550
AK292532.1 - 3 2474
BC001892.1 - 4 946 3534629
BC041843.1 - 5 2573 5269181
BC065040.1 - 4 935 6147639
BM875537.1 - 1 57

View Full Product Details