Gene Transcripts

Gene Symbol
PBXIP1
Entrez Gene ID
57326
Gene Name
PBX homeobox interacting protein 1
Gene Aliases
HPIP
Chromosome Location
Chr. 1: 154944080 - 154956163 on Build GRCh38
UniGene ID
Hs.505806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317734.1 NP_001304663.1 2 136
NM_020524.3 NP_065385.2 3 225
GenBank mRNA AF221521.1 - 3 174
AK022497.1 - 2 95
AK023219.1 - 3 166
AK300544.1 - 3 189
AK310388.1 - 4 654
AK315551.1 - 3 206
BC016852.1 - 3 153 3900461
HY108652.1 - 3 225

View Full Product Details