Gene Transcripts
- Gene Symbol
- C3orf14
- Entrez Gene ID
- 57415
- Gene Name
- chromosome 3 open reading frame 14
- Gene Aliases
- HT021
- Chromosome Location
- Chr. 3: 62318973 - 62336213 on Build GRCh38
- UniGene ID
- Hs.47166
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291941.1 |
NP_001278870.1
|
3, 4 |
264 |
|
NM_001291942.1 |
NP_001278871.1
|
2, 3 |
152 |
|
NM_001291943.1 |
NP_001278872.1
|
3, 4 |
264 |
|
NM_020685.4 |
NP_065736.1
|
3, 4 |
371 |
|
XM_017006931.1 |
XP_016862420.1
|
3, 4 |
747 |
|
GenBank mRNA |
AF236158.1 |
- |
3, 4 |
201 |
|
AK310431.1 |
- |
3, 4 |
342 |
|
AK313915.1 |
- |
3, 4 |
371 |
|
BC017772.2 |
- |
2, 3 |
181 |
4307022 |
BU568674.1 |
- |
|
106 |
|
DA615061.1 |
- |
3, 4 |
222 |
|
DA783189.1 |
- |
3, 4 |
264 |
|
View Full Product Details