Gene Transcripts

Gene Symbol
USP31
Entrez Gene ID
57478
Gene Name
ubiquitin specific peptidase 31
Gene Aliases
Chromosome Location
Chr. 16: 23061407 - 23149301 on Build GRCh38
UniGene ID
Hs.183817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020718.3 NP_065769.3 16 3043
XM_017023496.1 XP_016878985.1 15 2954
XM_017023497.1 XP_016878986.1 16 4569
XM_017023498.1 XP_016878987.1 15 2356
XM_017023499.1 XP_016878988.1 13 2160
GenBank mRNA AB033029.1 - 5 1168
AJ586135.2 - 16 3043
AK092790.1 - 2 1059
AK126447.1 - 3 1838
AK126752.1 - 16 2726
BX648357.1 - 7 1381

View Full Product Details