Gene Transcripts

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
Chromosome Location
Chr. 12: 68842200 - 68963240 on Build GRCh38
UniGene ID
Hs.654387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005502.2 NP_001005502.1 8 1000
NM_001874.4 NP_001865.1 8 1032
NM_198320.3 NP_938079.1 8 1018
GenBank mRNA AF368463.1 - 8 990
AK130146.1 - 5 608
AK313180.1 - 8 990
BC022276.1 - 8 990 4768771
DA833423.1 - 5 559
J04970.2 - 8 1032

View Full Product Details