Gene Transcripts
- Gene Symbol
- AKTIP
- Entrez Gene ID
- 64400
- Gene Name
- AKT interacting protein
- Gene Aliases
- FT1, FTS
- Chromosome Location
- Chr. 16: 53491028 - 53504411 on Build GRCh38
- UniGene ID
- Hs.380897
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012398.2 |
NP_001012398.1
|
2, 3 |
304 |
|
NM_001308325.1 |
NP_001295254.1
|
2, 3 |
275 |
|
NM_022476.3 |
NP_071921.1
|
2, 3 |
275 |
|
XM_005256095.4 |
XP_005256152.1
|
2, 3 |
624 |
|
XM_005256096.4 |
XP_005256153.1
|
2, 3 |
202 |
|
XM_005256097.4 |
XP_005256154.1
|
2, 3 |
236 |
|
XM_005256098.4 |
XP_005256155.1
|
2, 3 |
200 |
|
XM_017023564.1 |
XP_016879053.1
|
2, 3 |
624 |
|
XM_017023565.1 |
XP_016879054.1
|
2, 3 |
202 |
|
XM_017023566.1 |
XP_016879055.1
|
2, 3 |
200 |
|
GenBank mRNA |
AK023320.1 |
- |
2, 3 |
210 |
|
AK222743.1 |
- |
2, 3 |
207 |
|
AK303503.1 |
- |
2, 3 |
177 |
|
BC001134.2 |
- |
2, 3 |
223 |
2967019 |
BC095401.1 |
- |
2, 3 |
197 |
30334599 |
BI545474.1 |
- |
2, 3 |
272 |
|
CR457308.1 |
- |
1, 2 |
47 |
|
View Full Product Details