Gene Transcripts

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
Chromosome Location
Chr. 16: 53491028 - 53504411 on Build GRCh38
UniGene ID
Hs.380897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012398.2 NP_001012398.1 8 897
NM_001308325.1 NP_001295254.1 8 868
NM_022476.3 NP_071921.1 8 868
XM_005256095.4 XP_005256152.1 8 1217
XM_005256096.4 XP_005256153.1 8 795
XM_005256097.4 XP_005256154.1 8 829
XM_005256098.4 XP_005256155.1 8 793
XM_017023564.1 XP_016879053.1 8 1217
XM_017023565.1 XP_016879054.1 8 795
XM_017023566.1 XP_016879055.1 8 793
GenBank mRNA AK023320.1 - 8 803
AK222743.1 - 8 800
BC001134.2 - 8 816 2967019
CR457308.1 - 7 640

View Full Product Details