Gene Transcripts

Gene Symbol
STAG3L4
Entrez Gene ID
64940
Gene Name
stromal antigen 3-like 4 (pseudogene)
Gene Aliases
STAG3L4P
Chromosome Location
Chr. 7: 67302638 - 67321526 on Build GRCh38
UniGene ID
Hs.632013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040585.1 6 982
NR_040586.1 5 906
GenBank mRNA AK023257.1 - 5 900
AK308931.1 - 6 764
BC026058.2 - 5 872 3936696

View Full Product Details