Gene Transcripts

Gene Symbol
SOWAHC
Entrez Gene ID
65124
Gene Name
sosondowah ankyrin repeat domain family member C
Gene Aliases
ANKRD57, C2orf26
Chromosome Location
Chr. 2: 109614334 - 109618987 on Build GRCh38
UniGene ID
Hs.355455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023016.3 NP_075392.2 1 1550
GenBank mRNA AK025523.1 - 1 1519
BC036652.1 - 1 1574 5267818

View Full Product Details