Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 17: 44141902 - 44162477 on Build GRCh38
UniGene ID
Hs.437059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171251.2 NP_001164722.1 3 709
NM_001321310.1 NP_001308239.1 3 709
NM_001321311.1 NP_001308240.1 3 709
NM_024032.4 NP_076937.2 3 709
XM_011525187.1 XP_011523489.1 3 559
XM_011525188.1 XP_011523490.1 3 559
XM_011525189.1 XP_011523491.1 3 559
XM_011525190.1 XP_011523492.1 3 559
XM_011525191.2 XP_011523493.1 3 559
XM_011525192.1 XP_011523494.1 3 559
XM_011525193.1 XP_011523495.1 3 559
XM_011525194.1 XP_011523496.1 3 559
XM_011525195.1 XP_011523497.1 3 559
XM_017025031.1 XP_016880520.1 3 732
XM_017025032.1 XP_016880521.1 3 731
GenBank mRNA AK021656.1 - 736
AK291924.1 - 3 703
AL834295.1 - 3 653
BC000121.1 - 3 589 3352851
BI829278.1 - 3 705

View Full Product Details