Gene Transcripts

Gene Symbol
TANGO6
Entrez Gene ID
79613
Gene Name
transport and golgi organization 6 homolog
Gene Aliases
TMCO7
Chromosome Location
Chr. 16: 68843547 - 69085182 on Build GRCh38
UniGene ID
Hs.13526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024562.1 NP_078838.1 2, 3 748
XM_011523327.2 XP_011521629.1 2, 3 807
GenBank mRNA AB051533.1 - 2, 3 748
AK095333.1 - 1, 2 294

View Full Product Details