Gene Transcripts

Gene Symbol
UQCC2
Entrez Gene ID
84300
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Gene Aliases
C6orf125, Cbp6, M19, MNF1, bA6B20.2
Chromosome Location
Chr. 6: 33696761 - 33711751 on Build GRCh38
UniGene ID
Hs.732448
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032340.3 NP_115716.1 3 318
GenBank mRNA AF086234.1 - 3 240
AK311835.1 - 3 267
AY568085.1 - 3 294
BC006007.1 - 3 294 4284544
BI604191.1 - 3 268
DA123098.1 - 3 318

View Full Product Details