Gene Transcripts

Gene Symbol
MON1A
Entrez Gene ID
84315
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
SAND1
Chromosome Location
Chr. 3: 49908869 - 49930012 on Build GRCh38
UniGene ID
Hs.655014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142501.1 NP_001135973.1 3 1272
NM_032355.3 NP_115731.2 4 1758
XM_006713345.3 XP_006713408.1 4 1524
XM_011534160.1 XP_011532462.1 4 1387
GenBank mRNA AK074404.1 - 4 1568
AK092018.1 - 3 1071
AK315630.1 - 4 1612
BC006299.2 - 4 1570 4079816
BC009459.1 - 4 1527 3536968
BC047022.1 - 3 1022 5248424

View Full Product Details