Gene Transcripts

Gene Symbol
MICALCL
Entrez Gene ID
84953
Gene Name
MICAL C-terminal like
Gene Aliases
Ebitein1
Chromosome Location
Chr. 11: 12286900 - 12359144 on Build GRCh38
UniGene ID
Hs.128196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032867.2 NP_116256.2 9 2336
GenBank mRNA AK027872.1 - 7 1663

View Full Product Details