Gene Transcripts

Gene Symbol
BTBD6
Entrez Gene ID
90135
Gene Name
BTB domain containing 6
Gene Aliases
BDPL
Chromosome Location
Chr. 14: 105248542 - 105251093 on Build GRCh38
UniGene ID
Hs.7367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033271.2 NP_150374.2 5 952
XM_005268188.3 XP_005268245.1 4 1120
GenBank mRNA AF353674.1 - 4 816
BC042525.1 - 5 922 4824525

View Full Product Details