Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030771.1 | NP_110398.1 | 2 | 462 | |
NM_080654.2 | NP_542385.1 | 2 | 462 | ||
GenBank mRNA | AF155108.1 | - | 2 | 1088 | |
AF382034.1 | - | 2 | 351 | ||
AK308494.1 | - | 3 | 410 | ||
AK313358.1 | - | 2 | 429 | ||
AL555720.3 | - | 2 | 446 | ||
AY598334.1 | - | 2 | 408 | ||
BC008496.1 | - | 2 | 430 | 4276656 | |
BC029421.1 | - | 2 | 137 |