Gene Transcripts

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
Chromosome Location
Chr. 12: 88035491 - 88050160 on Build GRCh38
UniGene ID
Hs.591009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009894.2 NP_001009894.2 5 627
GenBank mRNA AK095477.1 - 5 627
AK311222.1 - 4 495
AL137488.1 - 4 2535
BC035136.2 - 4 464
BC058014.1 - 4 492
BC092422.1 - 5 569 30386540
BX537868.1 - 5 599
BX640968.1 - 5 602
BX648026.1 - 4 463
CR936776.1 - 5 616

View Full Product Details