Gene Transcripts

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
Chromosome Location
Chr. 2: 30610382 - 30809527 on Build GRCh38
UniGene ID
Hs.660911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144575.2 NP_653176.2 7, 8 953
XM_011533159.2 XP_011531461.1 7, 8 950
XM_011533160.2 XP_011531462.1 8, 9 1141
XM_011533161.2 XP_011531463.1 7, 8 950
XM_011533163.2 XP_011531465.1 7, 8 950
XM_017005264.1 XP_016860753.1 8, 9 1141
XM_017005265.1 XP_016860754.1 7, 8 950
XM_017005266.1 XP_016860755.1 7, 8 950
GenBank mRNA AK074418.1 - 8, 9 1020
AK290298.1 - 7, 8 954
AK308037.1 - 7, 8 930
BC117345.1 - 7, 8 914 40125896
BC117347.1 - 7, 8 914 40125898
BX640793.1 - 7, 8 953

View Full Product Details