Gene Transcripts

Gene Symbol
STRADA
Entrez Gene ID
92335
Gene Name
STE20-related kinase adaptor alpha
Gene Aliases
LYK5, NY-BR-96, PMSE, STRAD, Stlk
Chromosome Location
Chr. 17: 63702832 - 63741970 on Build GRCh38
UniGene ID
Hs.514402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003786.2 NP_001003786.1 4 480
NM_001003787.2 NP_001003787.1 6 591
NM_001003788.2 NP_001003788.1 5 562
NM_001165969.1 NP_001159441.1 4 504
NM_001165970.1 NP_001159442.1 4 459
NM_153335.5 NP_699166.2 4 480
XM_005257797.2 XP_005257854.1 6 442
XM_005257798.2 XP_005257855.1 4 404
XM_005257799.2 XP_005257856.1 5 413
XM_005257800.2 XP_005257857.1 6 466
XM_005257801.4 XP_005257858.1 6 466
XM_005257803.4 XP_005257860.1 6 466
XM_011525466.2 XP_011523768.1 6 466
XM_011525467.2 XP_011523769.1 6 493
XM_017025312.1 XP_016880801.1 4 355
XM_017025313.1 XP_016880802.1 5 437
XM_017025314.1 XP_016880803.1 4 404
GenBank mRNA AF308302.1 - 6 430
AK074771.1 - 4 346
AK075005.1 - 4 309
AK293160.1 - 4 367
AK301331.1 - 4 331
AL832407.1 - 5 375
AY290821.2 - 4 480
BC043641.1 - 5 353 3908187
BG339051.1 - 366

View Full Product Details