Gene Transcripts
- Gene Symbol
- B3GALT5-AS1
- Entrez Gene ID
- 114041
- Gene Name
- B3GALT5 antisense RNA 1
- Gene Aliases
- C21orf88
- Chromosome Location
- Chr. 21: 39597147 - 39612822 on Build GRCh38
- UniGene ID
- Hs.375120
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026542.1 |
|
3 |
1029 |
|
GenBank mRNA |
AF426266.1 |
- |
3 |
648 |
|
AK129590.1 |
- |
2 |
736 |
|
View Full Product Details