Gene Transcripts

Gene Symbol
OSBPL7
Entrez Gene ID
114881
Gene Name
oxysterol binding protein like 7
Gene Aliases
ORP7
Chromosome Location
Chr. 17: 47807367 - 47821822 on Build GRCh38
UniGene ID
Hs.463320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145798.2 NP_665741.1 22 2593
XM_017024122.1 XP_016879611.1 22 2615
XM_017024123.1 XP_016879612.1 22 2733
XM_017024124.1 XP_016879613.1 21 2580
XM_017024125.1 XP_016879614.1 12 1516
GenBank mRNA AB208886.1 - 10 3440
AF323729.1 - 22 2582
AF392446.1 - 22 2646
BC028010.1 - 14 1781
BC065482.1 - 22 2565 6081784

View Full Product Details