Gene Transcripts

Gene Symbol
ANKRD22
Entrez Gene ID
118932
Gene Name
ankyrin repeat domain 22
Gene Aliases
Chromosome Location
Chr. 10: 88819899 - 88851975 on Build GRCh38
UniGene ID
Hs.217484
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144590.2 NP_653191.2 5 787
GenBank mRNA AK313969.1 - 5 559
BC021671.1 - 5 574 3682099
BM785517.1 - 5 630

View Full Product Details