Gene Transcripts

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
Chromosome Location
Chr. 18: 13663347 - 13726592 on Build GRCh38
UniGene ID
Hs.13034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098801.1 NP_001092271.1 5 1144
NM_152352.3 NP_689565.2 4 1004
GenBank mRNA AK055618.1 - 5 1100
AK309506.1 - 2 391
AK309819.1 - 2 278
DA573992.1 - 2 392

View Full Product Details