Gene Transcripts

Gene Symbol
OR2B11
Entrez Gene ID
127623
Gene Name
olfactory receptor family 2 subfamily B member 11
Gene Aliases
Chromosome Location
Chr. 1: 247451029 - 247451982 on Build GRCh38
UniGene ID
Hs.446999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004492.1 NP_001004492.1 1 871
GenBank mRNA BC137181.1 - 1 895
BC137205.1 - 1 895 9021202

View Full Product Details