Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080476.4 | NP_536724.1 | 6 | 696 | |
XM_017027664.1 | XP_016883153.1 | 6 | 514 | ||
XM_017027665.1 | XP_016883154.1 | 6 | 514 | ||
GenBank mRNA | AB086842.1 | - | 6 | 502 | |
AK075507.1 | - | 6 | 521 | ||
AK301900.1 | - | 6 | 496 | ||
AK308627.1 | - | 5 | 443 | ||
AY339061.1 | - | 5 | 436 | ||
AY358816.1 | - | 6 | 526 | ||
AY422169.1 | - | 6 | 696 | ||
BC030512.1 | - | 6 | 507 | 4942673 |