Gene Transcripts

Gene Symbol
SAMD8
Entrez Gene ID
142891
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
HEL-177, SMSr
Chromosome Location
Chr. 10: 75099592 - 75182123 on Build GRCh38
UniGene ID
Hs.744986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174156.1 NP_001167627.1 2 463
NM_144660.2 NP_653261.1 2 519
XM_005269541.4 XP_005269598.1 2 654
XM_011539311.1 XP_011537613.1 2 582
XM_011539312.2 XP_011537614.1 2 512
XM_017015738.1 XP_016871227.1 2 519
XM_017015739.1 XP_016871228.1 2 547
GenBank mRNA AK057811.1 - 2 447
BC080593.1 - 2 463
EU668332.1 - 2 447
GQ472208.1 - 2 447

View Full Product Details