Gene Transcripts

Gene Symbol
LAYN
Entrez Gene ID
143903
Gene Name
layilin
Gene Aliases
Chromosome Location
Chr. 11: 111540280 - 111563464 on Build GRCh38
UniGene ID
Hs.503831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258390.1 NP_001245319.1 3 509
NM_001258391.1 NP_001245320.1 2 485
NM_001318799.1 NP_001305728.1 2 119
NM_178834.4 NP_849156.1 2 485
XM_006718769.3 XP_006718832.1 2 98
XM_006718770.3 XP_006718833.1 2 313
GenBank mRNA AK055539.1 - 2 486
AK055654.1 - 2 273
AK293317.1 - 2 316
AK296231.1 - 2 273
AK298165.1 - 2 255
AY358554.1 - 3 334
BC025407.1 - 2 278 4820414
DB006476.1 - 2 339
DC398135.1 - 2 119

View Full Product Details