Gene Transcripts

Gene Symbol
C4orf26
Entrez Gene ID
152816
Gene Name
chromosome 4 open reading frame 26
Gene Aliases
AI2A4
Chromosome Location
Chr. 4: 75554997 - 75565893 on Build GRCh38
UniGene ID
Hs.24510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257072.1 NP_001244001.1 1, 2 110
NM_178497.3 NP_848592.2 1, 2 110
NR_046429.1 3, 4 265
NR_046430.1 2, 3 246
GenBank mRNA AK074237.1 - 1, 2 110
AK172776.1 - 1, 2 110
BC117342.1 - 1, 2 110 40125893
R32248.1 - 1, 2 80

View Full Product Details