Gene Transcripts

Gene Symbol
C2orf27A
Entrez Gene ID
29798
Gene Name
chromosome 2 open reading frame 27A
Gene Aliases
C2orf27, C2orf27B
Chromosome Location
Chr. 2: 131722491 - 131767404 on Build GRCh38
UniGene ID
Hs.635289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013310.3 NP_037442.3 1 112
GenBank mRNA AF038169.1 - 1 112

View Full Product Details