Gene Transcripts

Gene Symbol
C2orf27A
Entrez Gene ID
29798
Gene Name
chromosome 2 open reading frame 27A
Gene Aliases
C2orf27, C2orf27B
Chromosome Location
Chr. 2: 131722491 - 131767404 on Build GRCh38
UniGene ID
Hs.635289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013310.3 NP_037442.3 2 249
GenBank mRNA AF038169.1 - 2 249
BC071972.1 - 1 43

View Full Product Details