Gene Transcripts

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
Chromosome Location
Chr. 19: 45178745 - 45202715 on Build GRCh38
UniGene ID
Hs.103902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212550.4 NP_997715.1 2 1212
GenBank mRNA AK130151.1 - 1 53
AK130186.1 - 1 491
AK296695.1 - 2 1210

View Full Product Details