Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
Chromosome Location
Chr. 9: 111686175 - 111795949 on Build GRCh38
UniGene ID
Hs.428209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080551.2 NP_001074020.2 14 2376
NM_173521.4 NP_775792.4 16 2383
NR_109816.1 18 2585
XM_006716974.2 XP_006717037.1 18 2569
XM_006716975.2 XP_006717038.1 17 2464
XM_011518302.2 XP_011516604.1 16 4378
XM_011518303.1 XP_011516605.1 16 2350
XM_011518306.1 XP_011516608.1 13 1970
XM_011518309.1 XP_011516611.1 18 2569
XM_017014340.1 XP_016869829.1 16 3187
XM_017014341.1 XP_016869830.1 18 2569
GenBank mRNA AK126055.1 - 14 2374
AL833535.1 - 16 2377
BC144680.1 - 18 2509

View Full Product Details