Gene Transcripts

Gene Symbol
OR56B4
Entrez Gene ID
196335
Gene Name
olfactory receptor family 56 subfamily B member 4
Gene Aliases
OR11-67
Chromosome Location
Chr. 11: 6107684 - 6108835 on Build GRCh38
UniGene ID
Hs.632154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005181.2 NP_001005181.1 1 617
GenBank mRNA BC136879.1 - 1 617
BC136880.1 - 1 617 9020871

View Full Product Details