Gene Transcripts

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
Chromosome Location
Chr. 17: 65635540 - 66192131 on Build GRCh38
UniGene ID
Hs.408676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037325.2 NP_001032402.1 3, 4 627
NM_001199165.2 NP_001186094.1 23, 24 2758
NM_001302891.1 NP_001289820.1 21, 22 2708
XM_005257119.4 XP_005257176.1 23, 24 2852
XM_005257125.3 XP_005257182.1 15, 16 1842
XM_005257126.4 XP_005257183.1 12, 13 1507
XM_006721740.2 XP_006721803.1 23, 24 2724
XM_011524461.2 XP_011522763.1 23, 24 2865
XM_011524462.2 XP_011522764.1 23, 24 2854
XM_011524463.2 XP_011522765.1 23, 24 2854
XM_011524464.2 XP_011522766.1 21, 22 2641
XM_011524465.2 XP_011522767.1 20, 21 2181
XM_011524466.2 XP_011522768.1 13, 14 1660
XM_011524467.2 XP_011522769.1 12, 13 1502
XM_017024302.1 XP_016879791.1 21, 22 2595
GenBank mRNA AF458591.1 - 13, 14 1665
AK097612.1 - 5, 6 726
AK098070.1 - 13, 14 1577
AK299075.1 - 23, 24 2709
BC029524.1 - 3, 4 627
BC131805.1 - 21, 22 2536 40119069
BX097629.1 - 3, 4 305

View Full Product Details