Gene Transcripts
- Gene Symbol
- AK9
- Entrez Gene ID
- 221264
- Gene Name
- adenylate kinase 9
- Gene Aliases
- AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
- Chromosome Location
- Chr. 6: 109492855 - 109691253 on Build GRCh38
- UniGene ID
- Hs.205144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145128.2 |
NP_001138600.2
|
9, 10 |
919 |
|
NM_145025.4 |
NP_659462.1
|
9, 10 |
919 |
|
XM_006715376.3 |
XP_006715439.2
|
10, 11 |
1784 |
|
XM_011535550.2 |
XP_011533852.1
|
10, 11 |
1784 |
|
XM_011535552.2 |
XP_011533854.1
|
9, 10 |
960 |
|
XM_011535553.2 |
XP_011533855.1
|
9, 10 |
1567 |
|
XM_011535554.2 |
XP_011533856.1
|
10, 11 |
1784 |
|
XM_011535555.2 |
XP_011533857.1
|
10, 11 |
1784 |
|
XM_011535556.2 |
XP_011533858.1
|
10, 11 |
1784 |
|
XM_011535559.2 |
XP_011533861.1
|
10, 11 |
1784 |
|
XM_017010384.1 |
XP_016865873.1
|
10, 11 |
1784 |
|
XM_017010385.1 |
XP_016865874.1
|
9, 10 |
952 |
|
XM_017010386.1 |
XP_016865875.1
|
10, 11 |
1784 |
|
XM_017010387.1 |
XP_016865876.1
|
4, 5 |
505 |
|
XM_017010388.1 |
XP_016865877.1
|
10, 11 |
1784 |
|
GenBank mRNA |
AK055538.1 |
- |
9, 10 |
896 |
|
AK315561.1 |
- |
9, 10 |
890 |
|
AL832162.1 |
- |
9, 10 |
898 |
|
BC022031.2 |
- |
9, 10 |
924 |
|
View Full Product Details